3-(4-Methoxy-phenoxy)-propylamine 95%

Lieferant: Apollo Scientific

Synonyme: 3-(4-Methoxyphenoxy)propan-1-amine

OR962630-1G
APOSOR962630-1GEA 144 EUR
APOSOR962630-1G
3-(4-Methoxy-phenoxy)-propylamine 95%
3-(4-Methoxy-phenoxy)-propylamine
Formel: C₁₀H₁₅NO₂
Molecular Weight: 181,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05053658
CAS-Nummer: 100841-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR