3-Pyrrolidin-1-ylbenzaldehyde 97%

Lieferant: Apollo Scientific

OR962684-1G OR962684-250MG
APOSOR962684-1GEA 270 EUR
APOSOR962684-1G APOSOR962684-250MG
3-Pyrrolidin-1-ylbenzaldehyde 97%
Formel: C₁₁H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD07772825
CAS-Nummer: 857283-89-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR