3-[1-(Dimethylamino)ethyl]phenol 97%

Lieferant: Apollo Scientific

Synonyme: 3-(1-(Dimethylamino)ethyl)phenol

OR962693-25G
APOSOR962693-25GEA 88.6 EUR
APOSOR962693-25G
3-[1-(Dimethylamino)ethyl]phenol 97%
3-[1-(Dimethylamino)ethyl]phenol
Formel: C₁₀H₁₅NO
Molecular Weight: 165,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08272688
CAS-Nummer: 105601-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR