tert-Butyl-5-bromo-1H-indazole-1-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-1H-indazole, N1-BOC protected, tert-Butyl 5-bromo-1H-indazole-1-carboxylate

OR9627-1G
APOSOR9627-1GEA 128 EUR
APOSOR9627-1G
tert-Butyl-5-bromo-1H-indazole-1-carboxylate
tert-Butyl-5-bromo-1H-indazole-1-carboxylate
Formel: C₁₂H₁₃BrN₂O₂
Molecular Weight: 297.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08435913
CAS-Nummer: 651780-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR