2-(2-Methoxyethoxy)benzoic acid 95%

Lieferant: Apollo Scientific

OR962706-1G OR962706-250MG OR962706-5G
APOSOR962706-1GEA 0 EUR
APOSOR962706-1G APOSOR962706-250MG APOSOR962706-5G
2-(2-Methoxyethoxy)benzoic acid 95%
2-(2-Methoxyethoxy)benzoic acid
Formel: C₁₀H₁₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08700372
CAS-Nummer: 98684-32-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR