2-(1-Pyrrolidinyl)benzoic acid 97%

Lieferant: Apollo Scientific

OR962734-1G OR962734-5G
APOSOR962734-1GEA 77 EUR
APOSOR962734-1G APOSOR962734-5G
2-(1-Pyrrolidinyl)benzoic acid 97%
2-(1-Pyrrolidinyl)benzoic acid
Formel: C₁₁H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09035985
CAS-Nummer: 78648-27-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR