3-(2-Methoxyethoxy)benzoic acid 95%

Lieferant: Apollo Scientific

OR962735-1G OR962735-250MG
APOSOR962735-1GEA 0 EUR
APOSOR962735-1G APOSOR962735-250MG
3-(2-Methoxyethoxy)benzoic acid 95%
3-(2-Methoxyethoxy)benzoic acid
Formel: C₁₀H₁₂O₄
Lagertemperatur: Kühlschrank
MDL: MFCD09036534
CAS-Nummer: 152808-60-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR