4-(Pyrrolidin-1-yl)benzamide 95%

Lieferant: Apollo Scientific

OR962767-1G
APOSOR962767-1GEA 364 EUR
APOSOR962767-1G
4-(Pyrrolidin-1-yl)benzamide 95%
4-(Pyrrolidin-1-yl)benzamide
Formel: C₁₁H₁₄N₂O
Molecular Weight: 190,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907634
CAS-Nummer: 183557-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR