2-Bromo-4-cyanothiazole 96%

Lieferant: Apollo Scientific

OR962783-5G
APOSOR962783-5GEA 165 EUR
APOSOR962783-5G
2-Bromo-4-cyanothiazole 96%
2-Bromo-4-cyanothiazole
Formel: C₄HBrN₂S
Molecular Weight: 189.0331 g/mol
Siedepunkt: 276 °C
Schmelzpunkt: 60…70 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09909650
CAS-Nummer: 848501-90-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR