2-(Ethylsulphonyl)aniline 98%

Lieferant: Apollo Scientific

OR962789-1G
APOSOR962789-1GEA 63.8 EUR
APOSOR962789-1G
2-(Ethylsulphonyl)aniline 98%
2-(Ethylsulphonyl)aniline
Formel: C₈H₁₁NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09945923
CAS-Nummer: 31596-87-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR