2-Azetidinecarboxylic acid, 1-methyl-, (2R)-
Lieferant: Apollo Scientific
OR962822-1G
APOSOR962822-1GEA
1110
EUR
APOSOR962822-1G
2-Azetidinecarboxylic acid, 1-methyl-, (2R)-
2-Azetidinecarboxylic acid, 1-methyl-, (2R)-
Formel:
C₅H₉NO₂ Molecular Weight: 115,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22397975 CAS-Nummer: 255883-21-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...