3-(2-Methyl-1H-imidazol-1-yl)benzonitrile 95%

Lieferant: Apollo Scientific

OR962846-1G
APOSOR962846-1GEA 231 EUR
APOSOR962846-1G
3-(2-Methyl-1H-imidazol-1-yl)benzonitrile 95%
3-(2-Methyl-1H-imidazol-1-yl)benzonitrile
Formel: C₁₁H₉N₃
Molecular Weight: 183,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11520464
CAS-Nummer: 167758-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR