3H-Imidazo[4,5-c]pyridine-6-carboxylic acid methyl ester 95%

Lieferant: Apollo Scientific

OR962867-100MG OR962867-1G OR962867-250MG
APOSOR962867-100MGEA 108 EUR
APOSOR962867-100MG APOSOR962867-1G APOSOR962867-250MG
3H-Imidazo[4,5-c]pyridine-6-carboxylic acid methyl ester 95%
3H-Imidazo[4,5-c]pyridine-6-carboxylic acid methyl ester
Formel: C₈H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11656494
CAS-Nummer: 82523-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR