2-Cyclopropyl-3-nitro-1H-pyrazole 98%

Lieferant: Apollo Scientific

Synonyme: 3-Cyclopropyl-4-nitro-1H-pyrazole

OR962920-1G
APOSOR962920-1GEA 69.6 EUR
APOSOR962920-1G
2-Cyclopropyl-3-nitro-1H-pyrazole 98%
2-Cyclopropyl-3-nitro-1H-pyrazole
Formel: C₆H₇N₃O₂
Molecular Weight: 153,139 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16249530
CAS-Nummer: 1249581-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR