1-Phenyl-2-pentanon 95%

Lieferant: Apollo Scientific

OR962924-1G OR962924-5G
APOSOR962924-1GEA 0 EUR
APOSOR962924-1G APOSOR962924-5G
1-Phenyl-2-pentanon 95%
1-Phenyl-2-pentanon
Formel: (C₆H₅)CH₂C(O)(CH₂)₂CH₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00027142
CAS-Nummer: 6683-92-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR