1-(2-Methoxyethoxy)-2-nitrobenzene 98%

Lieferant: Apollo Scientific

OR962926-100G OR962926-25G OR962926-5G
APOSOR962926-100GEA 0 EUR
APOSOR962926-100G APOSOR962926-25G APOSOR962926-5G
1-(2-Methoxyethoxy)-2-nitrobenzene 98%
1-(2-Methoxyethoxy)-2-nitrobenzene
Formel: C₉H₁₁NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD16877751
CAS-Nummer: 62819-95-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR