N-Methylpyridin-3-amine 98%

Lieferant: Apollo Scientific

Synonyme: 3-(Methylamino)pyridine

OR963110-25G
APOSOR963110-25GEA 112 EUR
APOSOR963110-25G
N-Methylpyridin-3-amine 98%
N-Methylpyridin-3-amine
Formel: C₆H₈N₂
Molecular Weight: 108,144 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01418130
CAS-Nummer: 18364-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR