Ac-Nle-OH 95%
Lieferant: Apollo Scientific
Synonyme:
N-Ac-Nle-OH, (S)-2-Acetamidohexanoic acid, (2S)-2-Acetamidohexanoic acid
OR963188-25G
OR963188-5G
APOSOR963188-25GEA
214
EUR
APOSOR963188-25G
APOSOR963188-5G
Ac-Nle-OH 95%
Ac-Nle-OH
Formel:
C₈H₁₅NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151961 CAS-Nummer: 15891-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...