2-(Chloromethyl)morpholine hydrochloride 97%

Lieferant: Apollo Scientific

OR963189-1G
APOSOR963189-1GEA 120 EUR
APOSOR963189-1G
2-(Chloromethyl)morpholine hydrochloride 97%
2-(Chloromethyl)morpholine hydrochloride
Formel: C₅H₁₀ClNO·ClH
Molecular Weight: 172,053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18711391
CAS-Nummer: 144053-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR