Ac-Nva-OH 98%
Lieferant: Apollo Scientific
OR963214-25G
OR963214-5G
OR963214-100G
APOSOR963214-25GEA
270
EUR
APOSOR963214-25G
APOSOR963214-5G
APOSOR963214-100G
Ac-Nva-OH 98%
Ac-Nva-OH
Formel:
C₇H₁₃NO₃ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
15891-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...