N-Formyl-L-valine 95%

Lieferant: Apollo Scientific

Synonyme: For-Val-OH

OR963260-1G OR963260-25G OR963260-5G
APOSOR963260-1GEA 111 EUR
APOSOR963260-1G APOSOR963260-25G APOSOR963260-5G
N-Formyl-L-valine 95%
N-Formyl-L-valine
Formel: C₆H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00021029
CAS-Nummer: 4289-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR