N-Boc-1,4-diaminobutane-HCl

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl (4-aminobutyl)carbamate hydrochloride, N-1-Boc-1,4-diaminobutane·HCl, BOC-1,4-Diaminobutane Hydrochloride

OR963265-25G
APOSOR963265-25GEA 114 EUR
APOSOR963265-25G
N-Boc-1,4-diaminobutane-HCl
N-Boc-1,4-diaminobutane-HCl
Formel: C₉H₂₁ClN₂O₂
Molecular Weight: 224.729 g/mol
Schmelzpunkt: 155…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02090802
CAS-Nummer: 33545-98-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR