Ac-DL-Pra-Oet 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-acetamidopent-4-ynoate

OR963272-1G OR963272-5G
APOSOR963272-1GEA 183 EUR
APOSOR963272-1G APOSOR963272-5G
Ac-DL-Pra-Oet 95%
Ac-DL-Pra-Oet
Formel: C₉H₁₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00237477
CAS-Nummer: 23235-05-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR