(R)-Methyl-2-amino-3-chloropropanoate hydrochloride 95%

Lieferant: Apollo Scientific

OR963306-1G
APOSOR963306-1GEA 0 EUR
APOSOR963306-1G
(R)-Methyl-2-amino-3-chloropropanoate hydrochloride 95%
(R)-Methyl-2-amino-3-chloropropanoate hydrochloride
Formel: C₄H₉Cl₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00270526
CAS-Nummer: 17136-54-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR