6-(2-Thienyl)-2-pyridinecarboxaldehyde 95%

Lieferant: Apollo Scientific

OR963394-1G
APOSOR963394-1GEA 255 EUR
APOSOR963394-1G
6-(2-Thienyl)-2-pyridinecarboxaldehyde 95%
6-(2-Thienyl)-2-pyridinecarboxaldehyde
Formel: C₁₀H₇NOS
Molecular Weight: 189,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410007
CAS-Nummer: 208111-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR