4-Iodo-1-isobutyl-1H-pyrazole ≥98%

Lieferant: Apollo Scientific

OR963402-5G
APOSOR963402-5GEA 59.6 EUR
APOSOR963402-5G
4-Iodo-1-isobutyl-1H-pyrazole ≥98%
4-Iodo-1-isobutyl-1H-pyrazole
Formel: C₇H₁₁IN₂
Molecular Weight: 250.08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11054044
CAS-Nummer: 918487-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR