6-(3-Thienyl)pyridine-2-carboxaldehyde 95%
Lieferant: Apollo Scientific
OR963406-1G
APOSOR963406-1GEA
284
EUR
APOSOR963406-1G
6-(3-Thienyl)pyridine-2-carboxaldehyde 95%
6-(3-Thienyl)pyridine-2-carboxaldehyde
Formel:
C₁₀H₇NOS Molecular Weight: 189,234 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06798135 CAS-Nummer: 834884-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...