Ethyl 2,2-dimethyl-3-oxopyrrolidine-1-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: 2,2-Dimethyl-3-oxo-pyrrolidine-1-carboxylic acid ethyl ester, Ethyl 2,2-dimethyl-3-oxopyrrolidine-1-carboxylate

OR963490-1G
APOSOR963490-1GEA 68.3 EUR
APOSOR963490-1G
Ethyl 2,2-dimethyl-3-oxopyrrolidine-1-carboxylate 95%
Ethyl-2,2-dimethyl-3-oxopyrrolidine-1-carboxylate
Formel: C₉H₁₅NO₃
Molecular Weight: 185.2200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09835297
CAS-Nummer: 106556-66-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR