2,3-Dimethoxyanilin 95%
Lieferant: Apollo Scientific
Synonyme:
(2,3-Dimethoxyphenyl)amine
OR963549-5G
APOSOR963549-5GEA
108
EUR
APOSOR963549-5G
2,3-Dimethoxyanilin 95%
2,3-Dimethoxyanilin
Formel:
C₈H₁₁NO₂ Molecular Weight: 153.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00180769 CAS-Nummer: 6299-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...