Boc-L-alaninal 95%

Lieferant: Apollo Scientific

Synonyme: N-tert-Butoxycarbonyl-L-alaninal, Boc-L-Ala-aldehyde, Boc-L-alaninal, Boc-Ala-aldehyde, (S)-(1-Oxopropan-2-yl)carbamic acid tert-Butyl ester, (S)-tert-Butyl (1-Oxopropan-2-yl)carbamate

OR963550-5G
APOSOR963550-5GEA 159 EUR
APOSOR963550-5G
Boc-L-alaninal 95%
Boc-L-alaninal
Formel: C₈H₁₅NO₃
Molecular Weight: 173.212 g/mol
Schmelzpunkt: 82…84 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00143786
CAS-Nummer: 79069-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR