Methyl-2-aminohexanoate hydrochloride 98%

Lieferant: Apollo Scientific

OR963597-100G OR963597-25G OR963597-5G
APOSOR963597-100GEA 0 EUR
APOSOR963597-100G APOSOR963597-25G APOSOR963597-5G
Methyl-2-aminohexanoate hydrochloride 98%
Methyl-2-aminohexanoate hydrochloride
Formel: C₇H₁₆ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00058296
CAS-Nummer: 77300-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR