Ethyl-6-methyl-2-phenylimidazo[1,2-a]pyridin-3-carboxylat

Lieferant: Apollo Scientific
Achtung

OR9636-250MG
APOSOR9636-250MGEA 232 EUR
APOSOR9636-250MG
Ethyl-6-methyl-2-phenylimidazo[1,2-a]pyridin-3-carboxylat
Ethyl-6-methyl-2-phenylimidazo[1,2-a]pyridin-3-carboxylat
Formel: C₁₇H₁₆N₂O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 127801-87-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR