2-Amino-3-methyl-3H-imidazole-4-carboxylic acid ethyl ester 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-amino-1-methyl-1H-imidazole-5-carboxylate

OR963687-1G
APOSOR963687-1GEA 136 EUR
APOSOR963687-1G
2-Amino-3-methyl-3H-imidazole-4-carboxylic acid ethyl ester 98%
2-Amino-3-methyl-3H-imidazole-4-carboxylic acid ethyl ester
Formel: C₇H₁₁N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09997799
CAS-Nummer: 177760-04-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR