(S)-Methyl-2-aminopentanoate hydrochloride 98%

Lieferant: Apollo Scientific

OR963720-100G OR963720-25G
APOSOR963720-100GEA 0 EUR
APOSOR963720-100G APOSOR963720-25G
(S)-Methyl-2-aminopentanoate hydrochloride 98%
(S)-Methyl-2-aminopentanoate hydrochloride
Formel: C₆H₁₄ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00190974
CAS-Nummer: 56558-30-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR