1-(2-Bromo-4-hydroxyphenyl)ethanone 98%

Lieferant: Apollo Scientific
Achtung

OR963788-1G OR963788-250MG OR963788-5G
APOSOR963788-1GEA 46.4 EUR
APOSOR963788-1G APOSOR963788-250MG APOSOR963788-5G
1-(2-Bromo-4-hydroxyphenyl)ethanone 98%
1-(2-Bromo-4-hydroxyphenyl)ethanone
Formel: C₈H₇BrO₂
Molecular Weight: 215.044 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03428552
CAS-Nummer: 61791-99-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR