2-Cyclopropylpyrimidine-5-carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-Cyclopropyl-pyrimidine-5-carboxylic acid

OR963924-1G
APOSOR963924-1GEA 178 EUR
APOSOR963924-1G
2-Cyclopropylpyrimidine-5-carboxylic acid 95%
2-Cyclopropylpyrimidine-5-carboxylic acid
Formel: C₈H₈N₂O₂
Molecular Weight: 164,160 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09054852
CAS-Nummer: 648423-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR