2-Methoxy-4-(2-nitroethyl)phenol 95%

Lieferant: Apollo Scientific

OR963988-1G
APOSOR963988-1GEA 307 EUR
APOSOR963988-1G
2-Methoxy-4-(2-nitroethyl)phenol 95%
2-Methoxy-4-(2-nitroethyl)phenol
Formel: C₉H₁₁NO₄
Molecular Weight: 197,188 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03410699
CAS-Nummer: 528594-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR