4-Phenoxyphenacyl bromide 90%
Lieferant: Apollo Scientific
|
Gefahr
|
OR9640-1G
OR9640-250MG
APOSOR9640-1GEA
401
EUR
APOSOR9640-1G
APOSOR9640-250MG
4-Phenoxyphenacyl bromide 90%
4-Phenoxyphenacyl bromide
Formel:
C₁₄H₁₁BrO₂ Schmelzpunkt: 49…50 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01419855 CAS-Nummer: 28179-33-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...