2-Methyl-5-nitro-1H-pyrrolo[2,3-b]pyridine 95%

Lieferant: Apollo Scientific

OR964024-1G
APOSOR964024-1GEA 145 EUR
APOSOR964024-1G
2-Methyl-5-nitro-1H-pyrrolo[2,3-b]pyridine 95%
2-Methyl-5-nitro-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₇N₃O₂
Molecular Weight: 177,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12962919
CAS-Nummer: 1190322-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR