(R)-(-)-Methylmandelat 98%

Lieferant: Apollo Scientific

Synonyme: D(-)-Methylmandelat, Methyl-D(-)-glykolat, D(-)-2-Hydroxy-2-phenylessigsäuremethylester, D(-)-Mandelsäuremethylester, Methyl-(R)-(-)-glykolat, (R)-(-)-Methylglykolat, (R)-(-)-Mandelsäuremethylester, (R)-(-)-2-Hydroxy-2-phenylethansäuremethylester, (R)-(-)-2-Hydroxy-2-phenylessigsäuremethylester, D(-)-Methylglykolat, D-2-Hydroxy-2-phenylethansäuremethylester

OR964083-100G
APOSOR964083-100GEA 84.1 EUR
APOSOR964083-100G
(R)-(-)-Methylmandelat 98%
(R)-(-)-Methylmandelat
Formel: C₉H₁₀O₃
Molecular Weight: 166.18 g/mol
Siedepunkt: 267
Lagertemperatur: Raumtemperatur
MDL: MFCD00064247
CAS-Nummer: 20698-91-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR