6-Methoxy-1,2,3,4-tetrahydrochinolin 95%

Lieferant: Apollo Scientific

OR964112-5G
APOSOR964112-5GEA 143 EUR
APOSOR964112-5G
6-Methoxy-1,2,3,4-tetrahydrochinolin 95%
6-Methoxy-1,2,3,4-tetrahydrochinolin
Formel: C₁₀H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00090494
CAS-Nummer: 120-15-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR