3-(1H-Imidazol-2-yl)pyridine 95%

Lieferant: Apollo Scientific

OR964166-1G
APOSOR964166-1GEA 58 EUR
APOSOR964166-1G
3-(1H-Imidazol-2-yl)pyridine 95%
3-(1H-Imidazol-2-yl)pyridine
Formel: C₈H₇N₃
Molecular Weight: 145.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458369
CAS-Nummer: 13570-00-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR