3-Phenoxybenzoyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR9642-1G
OR9642-250MG
APOSOR9642-1GEA
0
EUR
APOSOR9642-1G
APOSOR9642-250MG
3-Phenoxybenzoyl chloride 97%
3-Phenoxybenzoyl chloride
Formel:
C₁₃H₉ClO₂ Siedepunkt: 174…175 °C (12 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424712 CAS-Nummer: 3586-15-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...