3-(3-Methoxyphenyl)propionsäure 97%

Lieferant: Apollo Scientific

Synonyme: 3-(3-Methoxyphenyl)propansäure

OR964229-25G
APOSOR964229-25GEA 111 EUR
APOSOR964229-25G
3-(3-Methoxyphenyl)propionsäure 97%
3-(3-Methoxyphenyl)propionsäure
Formel: C₁₀H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00014027
CAS-Nummer: 10516-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR