3-(4-Chlorophenyl)azetidine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR964271-250MG
APOSOR964271-250MGEA 1130 EUR
APOSOR964271-250MG
3-(4-Chlorophenyl)azetidine hydrochloride 95%
3-(4-Chlorophenyl)azetidine hydrochloride
Formel: C₉H₁₀ClN·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08460697
CAS-Nummer: 7606-31-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR