D-Leucine ethyl ester hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: H-D-Leu-OEt.HCl, H-D-Leu-OEt·HCl

OR964283-25G
APOSOR964283-25GEA 58 EUR
APOSOR964283-25G
D-Leucine ethyl ester hydrochloride 95%
D-Leucine ethyl ester hydrochloride
Formel: C₈H₁₇NO₂·HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00190936
CAS-Nummer: 73913-65-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR