5-Methyl-2-phenyl-3-furoyl chloride 97%

Lieferant: Apollo Scientific

Synonyme: 5-Methyl-2-phenylfuran-3-carbonyl chloride

OR9643-1G
APOSOR9643-1GEA 346 EUR
APOSOR9643-1G
5-Methyl-2-phenyl-3-furoyl chloride 97%
5-Methyl-2-phenyl-3-furoyl chloride
Formel: C₁₂H₉ClO₂
Molecular Weight: 220,652 g/mol
Siedepunkt: 120…125 °C (0,5 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD08435904
CAS-Nummer: 183210-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR