3,3-Dimethylindolin-2-one 97%
Lieferant: Apollo Scientific
OR964406-5G
APOSOR964406-5GEA
140
EUR
APOSOR964406-5G
3,3-Dimethylindolin-2-one 97%
3,3-Dimethylindolin-2-one
Formel:
C10H11NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00158549 CAS-Nummer: 19155-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...