3,4-Dichloroisothiazole-5-carbonitrile 95%

Lieferant: Apollo Scientific

OR964430-1G
APOSOR964430-1GEA 318 EUR
APOSOR964430-1G
3,4-Dichloroisothiazole-5-carbonitrile 95%
3,4-Dichloroisothiazole-5-carbonitrile
Formel: C₄Cl₂N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09743745
CAS-Nummer: 18480-52-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR