2-(Ethoxycarbonyl)isonicotinic acid 95%

Lieferant: Apollo Scientific

OR964463-1G
APOSOR964463-1GEA 647 EUR
APOSOR964463-1G
2-(Ethoxycarbonyl)isonicotinic acid 95%
2-(Ethoxycarbonyl)isonicotinic acid
Formel: C₉H₉NO₄
Molecular Weight: 195,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08064021
CAS-Nummer: 142074-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR